BRAND® Inserts 2 in 1 for Cell Culture, BrandTech® Scientific, Inc.
Catalog #: 76337-190
Supplier: Brandtech
Specifications
- Culture area:0.6 cm²
- Membrane Material:Polyethylene terephthalate (PET)
- Pore size:8.0 µm
- Packaging:48, Multi-pack blister
- Working Volume:150 - 400 µl
- Membrane Diameter:8.8 mm
Specifications
About this item
BRAND® insert 2 in 1 offers unlimited flexibility for 2D and 3D cell culture. Inserts can be used as a free standing or hanging insert for co-culture, air-lift cultures, migration assays and more.
- cellGrade™ plus surface for enhanced cell attachment
- PC or PET membrane
- Available in 0.4 µm, 1 µm, 3 µm, or 8 µm pore sizes
- Compatible with all common 6-, 12-, or 24-well plates and tissue culture dishes
Common applications include co-culture, trans-migration, chemotaxis, transport/barrier analysis (TEER), cytotoxicity and air-lift culture assays.
Pyrogen and endotoxin free according to LAL test, detection limit 0.01 EU/ml.
Certifications: Manufactured in a Class 8 cleanroom, in accordance with ISO 14644-1. Sterile according to ISO 11137, SAL 10-⁶. Non-cytoxic according to ISO 10993-5.
Packaging: Inserts are available individually packed or in multi-packs.
Caution: These products can expose you to chemicals including α-Methyl styrene (alpha-Methylstyrene), which is known to cause cancer, and Di-n-butyl Phthalate (DBP), which is known to cause birth defects or other reproductive harm.
Customers Who Bought This Also Bought