VWR® Ratchet Headgear with Face Shield
Catalog #: 76336-850
Supplier: VWR International
Specifications
- Description:Headgear and shield
- Thickness:2.0 mm
- Cat. no.:76336-850
Specifications
About this item
The VWR® Ratchet Headgear with Face Shield provides full face protection from debris and splash hazards. Ideal for use in many environments ranging from the lab to industrial applications.
- Lightweight design
- Ratchet headgear features padded headband for better fit and comfort
- Face shield features universal slot design compatible with many different headgear styles
- Fits comfortably with respirator or eyewear
- Impact-resistant, scratch-resistant polycarbonate shield
Face shields should be worn with ANSI approved eyewear.
Certifications: ANSI Z87.1 certified
Customers Who Bought This Also Bought