Specifications
- Dimensions:24×24"
- Description:Steri-Wrap® II Single Use Non-Woven Wraps
- Cat. no.:76334-062
Specifications
About this item
Single use non-woven fabric sterilization wrap offers greater drapeability than standard wraps.
- Cellulose based wrap for steam and EO sterilization
- Efficient microbial barrier, high-level of protection against fluids and particles
- Highly permeable meaning no wet packs
- Variety of strength of fabric to meet your needs
Steri-Wrap® is cellulose based sterilization wrap available in three grades for keeping packages safe and secure during the sterilization process. Steri-Wrap® II is light weight wrap designed for great drapeability.
Steri-Wrap® is designed to be virtually lint free with tightly anchored fibers. Wrap is reversible and features excellent liquid repellency. Steri-Wrap® can be used in steam or ethylene oxide sterilization.