Water Bath Protective Agent, polyclear MIX 30 PLUS, PolyScience
Catalog #: 76333-932
Supplier: Polyscience
CAS Number:
Specifications
- Description:Water Bath Protective Agent, polyclear MIX 30 PLUS
- Size:1.9 l (0.5 gal.)
- Temperature Range:5...90 °C (41..194 °F)
Specifications
About this item
General purpose fluid that prevents algae growth and premature rust formation. Ideal for use in heating baths, circulators, water baths and shaking water baths.
- For routine applications above 5 °C
- Easy to use, no dilution required
Customers Who Bought This Also Bought