Specifications
- Cap Type:Open Top
- Capacity:38.5 ml
- Dimensions:25×89 mm
- Format:Round bottom
- Material:Ultra-Clear
- Compatibility:Type 70 Ti, Type 50.2 Ti, SW 32 Ti, SW 28, JS-24.38
Specifications
About this item
These tubes help protect your sample from degradation and contamination while solving complex biological problems, investigating the causes of diseases or finding potential new cures. They provide sample protection when it matters most.
- Maximize consistency
- Prevent yield loss
- Ready-to-go, right out of the bag
Certified free: These tubes exhibit no detectable DNA, DNase, RNase, PCR inhibitors, or endotoxins. (Based on sample results below detectable limits.)
• Ideal for genomics, exosomes, and proteomics workflows
• Prevents sample degradation and contamination
• Double-bagged to prevent contamination during clean room transfer
• Improve QC data quality
Certified free + Sterile: Beckman sterile ultracentrifuge tubes are sterilized via ethylene oxide.
• Ideal for protein prep and virus purification workflows
• Includes all advantages of being certified free
• Pre-sterilized ready-to-use for improved workflow efficiency
• Eliminate need for developing sterilization protocolol
• Packaged in workflow based quantities
• Sterilized and packaged compliant with ISO 11135 and 11607
Unique applications and workflows have varying requirements for the presence of viable and nonviable biologic materials. Samples may require consumables verified as certified free, sterile, or both. Choosing the right tube, provides your sample or experiment the best possible outcome.
Certified free: Genomics, DNA/RNA sequencing, exosomes and proteomics.
Certified free + Sterile: Microbiology, gene therapy, biotherapeutics, molecular biology, cell culture, plant/animal derived products and algae.