PIPET CAL ISO17025 4R/3V AL
Catalog #: CA76332-778
Supplier: COVARIANCE SV
Customers Who Bought This Also Bought
n-Heptane ≥99% (n-heptane) [≥99.9% (saturated C7 hydrocarbons)], OmniSolv®, Supelco®
Supplier: MilliporeSigma
$971.75 to $3,463.97
Extran® MN 01 Powdered Detergent for Manual Washing, MilliporeSigma
Supplier: MilliporeSigma
$214.10 to $1,039.32
Hydrogen peroxide 30% stabilized, CMOS for microelectronic use, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$218.47 to $218.47
Acetic acid glacial ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® ACS aldehyde-free, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$330.55 to $1,952.15
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) solution 10%, sterile
Supplier: Teknova
$386.34
Hydrogen peroxide 35% (w/w) in aqueous solution stabilized
Supplier: Thermo Scientific Chemicals
$311.01