PIPET CAL M12 ISO8655 10R/3V AL
Catalog #: CA76332-774
Supplier: COVARIANCE SV
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Ocean Pacific® Ninja™ Powder-Free Nitrile Medical Examination Gloves, Medicom
Supplier: Medicom
$53.49 to $523.36
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$39.55
Diethyl ether, anhydrous ≥99.0% (by GC) stabilized, GR ACS, Supelco®
Supplier: MilliporeSigma
$602.62 to $3,475.79
Sorensen's Phosphate Buffer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$86.00