SpotSee ShockWatch Temperature Indicators
Catalog # 76330-004
Supplier: Cold Chain Technologies
Specifications
- Temperature Range:0 °C/32 °F (48 hrs.)
- Description:WarmMark™ Indicator
- Cat. no.:76330-004
Specifications
About this item
ColdMark and WarmMark time–temperature indicators provide a cost-effective tool for monitoring the temperature of your package. They are single-use devices that provide accurate, irreversible evidence of a temperature excursion. With the information you gather from these indicators, you can make smarter decisions across your cold chain.
- Aids in compliance with regulatory guidelines
- Delivers irreversible evidence of exposure to unacceptable temperature conditions
- Assists in verifying the adequacy of the cold chain packaging
- Enables easy accept/reject decisions to be made
- Provides cost–effective solution for last mile monitoring
Temperature indicators introduce accountability measures into the shipping and handling stages of the temperature sensitive cold chain. In the event that a temperature excursion occurs, indicators give you the data needed to improve the integrity of the cold chain.
Frequently Bought Together