Specifications
- Capacity:3000 ml
- Color:Amber
- Description:24-Hour Urine Collection Container, Amber
Specifications
About this item
Ideal for collecting specimens over a 24-hour period, these leak resistant, wide mouth bottles facilitate the collection, transport and retrieval of specimens.
- Special gasketed screw cap enhances the container's leak resistance
- Containers are graduated from 100 to 3000 cc (ml) in 100 cc increments
- Latex-Free
- Compatible with calculi strainer (76329-592)
Available in amber or translucent white. The amber container is ideal for transporting light sensitive specimens.