Flash Spherical Diol Column, Particle size: 40-60um, Pore size: 100A, Silica amount: 220g
Catalog #: 76329-168
Supplier: Agela Technologies
Flash Spherical Diol Column, Particle size: 40-60um, Pore size: 100A, Silica amount: 220g
Catalog #: 76329-168
Supplier: Agela Technologies
Customers Who Bought This Also Bought
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
214,30 $ to 2 349,04 $
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
194,14 $ to 694,12 $
Sorensen's Phosphate Buffer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
86,00 $