Diatomite 96-Deep-Well Plate, (Ph=9) Sample Loading 500uL, 100-200 Mesh
Catalog #: 76327-604
Supplier: Agela Technologies
Diatomite 96-Deep-Well Plate, (Ph=9) Sample Loading 500uL, 100-200 Mesh
Catalog #: 76327-604
Supplier: Agela Technologies
Customers Who Bought This Also Bought
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Supplier: Air-Tite
$105.11 to $2,771.71
Nickel, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, VWR Chemicals BDH®
Supplier: VWR International
$110.90
Hydrogen peroxide 35% (w/w) in aqueous solution stabilized
Supplier: Thermo Scientific Chemicals
$311.01
J.T.Baker® Cell Lysis Solution, Ultrapure Bioreagent
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$287.56
WEBCOL® Alcohol Preps, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$15.59 to $18.12