Order Entry
ContactUsLinkComponent
Buffer Pouch Ph Calib Assorted 20Ml PK20
Oakton® pH Buffer Pouches, Cole-Parmer
Catalog #: 76325-404
Supplier:  Argos Technologies
CAS Number:  
Buffer Pouch Ph Calib Assorted 20Ml PK20
Oakton® pH Buffer Pouches, Cole-Parmer
Catalog #: 76325-404
Supplier:  Argos Technologies
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • pH:
    4.01/7.00/10.00
  • Color:
    Clear
  • Description:
    pH Calibration Buffer Pouches
  • Size:
    20× 20 ml pouches

Specifications

About this item

Calibrate your pH meter in the lab or in the field with convenient, portable, and highly accurate pH Singles

  • No unwieldy bottles to carry or clean
  • Economical pouches are accurate and convenient
  • Buffers are NIST-traceable and offer ±0.01 accuracy at 77 °F (25 °C)

The pouches are airtight for maximum freshness and unquestionable reliability.

Ordering information: Includes 20 single-use pouches in a dispensing box, with NIST-traceable calibration certificate.
Customers Who Bought This Also Bought