Oakton® pH Buffer Solutions, Cole-Parmer
Catalog #: 76325-386
Supplier: Argos Technologies
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- pH:7.00
- Color:Yellow
- Description:pH buffer solution
- Size:4 L
Specifications
About this item
Ideal for laboratory and field analysis
- pH buffers are traceable to NIST standard reference materials
- Buffer solution lets you calibrate pH instruments to ensure accuracy
- If unopened, shelf life is 2 years
- pH Buffer Solutions have pH/temperature tables printed on the labels for accurate calibrations (not on 60-mL, but included as insert)
- Manufactured under ISO 9000 quality guidelines
- If unopened, shelf life is 2 years; once opened, use pH 4 and pH 7 within 90 days, pH 10 within 30 days
Packaging: Plastic bottles.
Caution: Store buffers away from direct sunlight.
Customers Who Bought This Also Bought