SP Wilmad-LabGlass ProCulture® Fernbach Shaker Flasks, SP Industries
Catalog #: 76323-672
Supplier: SP Industries
Specifications
- Capacity:2800 ml
- O.D.:200 mm
- Description:Side baffles, beaded rim
- Height:230 mm
- Cat. no.:76323-672
- Stopper No.:13
- Thread:—
Specifications
About this item
Commonly used in serum production and ideal for use in organism cultures that require a large surface area to volume ratio, these flasks are available with a beaded rim, GL 45 screw thread, or Delong style neck for accommodating various style stoppers and plugs.
- Delong style neck reduces splashing to the cap, when on shaker
- Beaded rim style provides increased durability to the flask
- Baffles, spaced at 120°, allow for increased oxygen transfer to the Culture medium when used in conjunction with a shaker
- Autoclavable
Type 1, Class A borosilicate glass construction. Environmentally sustainable when compared to plastic.
Caution: Do not place on direct heat source
Customers Who Bought This Also Bought