Order Entry
ContactUsLinkComponent
Scissors Dissecting Student Grd Ss 4.5In
Student Dissection Scissors
Catalog #: 470315-162
Scissors Dissecting Student Grd Ss 4.5In
Student Dissection Scissors
Catalog #: 470315-162

Specifications

  • Description:
    Student Scissors
  • Material:
    Stainless Steel
  • Cat. no.:
    470315-162
  • Length:
    115 mm (4.5")
  • Blade Type:
    Sharp/Sharp

Specifications

About this item

Student grade.

  • Stainless steel construction
  • Sharp /sharp point configuration

Easier for students to use, these scissors are a durable stainless steel construction. Unlike a surgical style scissor, these have a gap between the handles when closed giving a shorter travel length for students cutting closer to the pivot point.

Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Customers Who Bought This Also Bought