Camera Adapters
Catalog #: 470314-906
Supplier: MOTIC INSTRUMENTS USA, INC SE
Specifications
- Description:C-mount adapter for 1/3" chip sensor, 0.35×
- Cat. no.:470314-906
- For Use With:Compatible cameras: Moticam 2, Moticam ×, Moticam ×2, Moticam 1080, Moticam BTU 10
Specifications
About this item
C-mount camera adapter allows Moticams to connect with Motic trinocular microscopes.
Customers Who Bought This Also Bought