Media/Packaging Bottles, PETG, Square, TriForest
Catalog #: 76320-390
Supplier: TRIFOREST ENTERPRISES INC
Specifications
- Cap Type:Standard Cap
- Capacity:125 ml
- Sterility:Sterile
- Description:PETG Square Media/Packaging Bottles
- Cat. no.:76320-390
- Packaging:24/Tray, 96/Case
Specifications
About this item
TriForest Square PETG bottles are ideal for storage of media, reagents, and buffers.
- Thick-walled and durable; shatter resistant
- Excellent gas barrier properties; reduced permeability to CO2/O2
- Sterility of 10⁻⁶ SAL
- Molded graduations for approximate volume reference
- Compliant with USP Class VI guidelines
- Non-pyrogenic, nuclease-free, and non-cytotoxic
- Bottle and cap material are Certified Animal Derived Component-Free
- Two litre bottles have a unique design with molded-in hand/robotic grips
Sterile polyethylene terephthalate co-polyester bottles are available with or without high density polyethylene caps in shrink wrapped trays. These heavy-duty, break-resistant bottles are designed for manual and automated fill lines.
Bottles are supplied shrink wrapped in polymer trays to eliminate static adherence to particulate matters. Double poly-bagged for unwrapping in cleanroom operations to maintain the utmost integrity of cleanroom based sterile production/filling/packaging requirements.
Bottles are available with standard cap (PGCXXXXMS-C), tamper evident cap (PGCXXXXMS-A), or without cap (PGCXXXXMS-N) for use with automated fill lines.
Customers Who Bought This Also Bought