VWR® Sharps Containers with Flip Up Lids
Catalog #: 76319-814
Supplier: VWR International
Specifications
- Capacity:2 gal.
- Color:Red
- Cat. no.:76319-814
- L×W×H:10.2×7×10.1"
Specifications
About this item
Containers provide for the safe disposal of sharps and biohazard waste. The flip up lid design is perfect for busy professional in settings such as laboratories, hospitals, dental, or physician offices requiring one-handed access to open and close sharps containers.
- Flip Up lid is intuitive to use, allowing one-handed operation
- Designed for use in high activity environments
- Translucent colors with lead-free pigments containing no heavy metals
- Constructed of puncture and impact resistant material
- Safe to autoclave or incinerate
- Stackable design
Choose from one of five sharps containers with flip up lid designs. Lightweight and transportable, the containers are ideal for any professional that needs an easy yet safe way to dispose of biohazardous waste. Quick access flip up lid lets users drop in waste and push down the lid with one hand to a 'temporary close' position for storage between use. Safe to autoclave or incinerate.
Certifications: Meets or exceeds FDA standards including OSHA and EPA requirements for local biohazard waste collection.
Customers Who Bought This Also Bought