VWR® Sharps Containers with Sliding Lid
Catalog #: 76319-796
Supplier: VWR International
Specifications
- Capacity:1 gal.
- Dimensions:10.2L×7W×5.6"H
- Color:Red
- Cat. no.:76319-796
Specifications
About this item
Containers provide for the safe disposal of sharps and bio-hazard waste. The design is perfect for busy professional settings such as laboratories, hospitals, dental, and physician offices requiring one-handed access to open sharps containers.
- Slide to close lid is intuitive to use, allowing one-handed operation
- Designed for use in high activity environments
- Slide door has both a temporary and permanent close position
- Translucent colors with lead-free pigments containing no heavy metals
- Constructed of puncture and impact resistant material
- Safe to autoclave or incinerate
- Stackable design
- Lightweight and transportable
Choose from one of four sharps containers with sliding lid designs. Quick access rotating door on the lid lets users drop in waste and move the lid to a 'temporary close' position for storage between use.
Certifications: Reviewed and approved directly by the FDA; Meets or exceeds FDA standards including OSHA and EPA requirements for local biohazard waste collection.
Customers Who Bought This Also Bought