VWR® PCR Tubes and Caps
Catalog #: 76318-804
Supplier: VWR International
Specifications
- Cap Type:Flat, Optically-Clear
- Capacity:0.2 ml
- Format:Tubes
- Color:Natural
- Cat. no.:76318-804
Specifications
About this item
Ideal for all PCR protocols, these 0.2 ml tubes and caps are made from polypropylene and are compatible with most popular thermal cycler blocks.
- Autoclavable
- Chemically inert
They feature smooth, thin, uniform walls for accurate thermal transfer. Certified free of RNase, DNase, and human DNA (lot-by-lot certificates available). Tubes are available with bubble-top or flat-top caps, both of which fit precisely to ensure a positive, contamination-free environment.
Customers Who Bought This Also Bought