Order Entry
ContactUsLinkComponent
Buchner Funnel Porcelain 60/18Mm Cs5
Porcelain Buchner Funnels
Catalog #: 470313-470
Supplier:  GSC International
Buchner Funnel Porcelain 60/18Mm Cs5
Porcelain Buchner Funnels
Catalog #: 470313-470
Supplier:  GSC International

Specifications

  • Cat. no.:
    470313-470
  • Top Diameter:
    60 mm
  • Stem Diameter:
    18 mm

Specifications

About this item

Cost-efficient, high quality Buchner Funnels.

  • Three sizes available
  • Fragile

These Buchner Funnels are made of a high-quality, laboratory-grade porcelain and have uniform wall thickness and dense materials to make a product with stable mechanical strength and little permeability. When properly used, the glazed pieces can withstand temperatures of 1050 °C and the unglazed pieces can withstand temperatures up to 1350 °C.

Customers Who Bought This Also Bought