Bottles, Boston Round, Amber / Clear
Catalog #: 470313-456
Supplier: GSC International
Specifications
- Capacity:2 oz.
- Color:Amber
- Cat. no.:470313-456
Specifications
About this item
The amber bottles with dropper assembly provide excellent UV protection for the content and the dropper allows for dispensing a small amount of liquid.
- Amber colored glass provides UV protection
- Provided with a dropper assembly or screw cap
- Pipette is made of glass
- Cap is made of plastic
- Bulb is made of rubber
Amber flint glass boston round bottles are available in multiple sizes. Sold with a dropper assembly or a cap.
Packaging: Sold in packs of 12.
Customers Who Bought This Also Bought