Glass Petri Dishes
Catalog #: 470313-348
Supplier: GSC International
Specifications
- Ø:100 mm
- Description:Petri Dish
- Cat. no.:470313-348
Specifications
About this item
These glass petri dishes are two-piece (lid and dish) and are packed individually.
- Covered dishes are not airtight
- Transparent and reusable
- Resistant to most acids, bases and alcohols
- Not autoclavable
Customers Who Bought This Also Bought