PTFE to Ace-Thred™ to Tubing Compression Fittings, Ace Glass Incorporated
Catalog #: 76318-554
Supplier: Ace Glass
Specifications
- O.D.:3/8"
- Cat. no.:76318-554
- Thread:#11 (ace-thread)
Specifications
About this item
PTFE fitting joins Ace-Thred™ glassware to tubing, via a compression fitting.
- Made completely of PTFE
- Fitting is composed of two PTFE adapters: (1) Ace-Thred to female NPT and (1) male NPT to tube compression fitting
- The two parts may be used in conjunction with each other, or as separate adapters for differing applications
Customers Who Bought This Also Bought