Ferrule Gripper Sets, Ace Glass Incorporated
Catalog #: 76318-538
Supplier: Ace Glass
Specifications
- Description:Ferrule/Gripper Set
- Size:1/4"
- Cat. no.:76318-538
Specifications
About this item
Replacement ferrule/gripper sets for ACE PTFE tube compression fittings.
Delivery: Each set consists of one each PTFE ferrule and PVDF gripper.
Customers Who Bought This Also Bought