Order Entry
ContactUsLinkComponent
VWR Uv Horizontal System Mini L 12X14Cm
VWR® Horizontal MINI L Gel Electrophoresis System
Catalog #: 76314-778
Supplier:  VWR International
VWR Uv Horizontal System Mini L 12X14Cm
VWR® Horizontal MINI L Gel Electrophoresis System
Catalog #: 76314-778
Supplier:  VWR International

Specifications

  • Gel size (W×L):
    12×14 cm
  • Description:
    VWR® MINI L Horizontal Gel System
  • No. of Samples:
    8 - 100
  • Cat. no.:
    76314-778
  • L×W×H:
    245×180×95 mm
  • Buffer Volume:
    800 ml

Specifications

About this item

The MINI L is the largest of the 'Mini-Gel' range is supplied with a four slot UVT gel tray capable of running 100 samples.

There is also a 12 slot tray available. There is an expansive range of standard and MC-compatible loading combs, making this versatile system appropriate for both routine and small separation electrophoresis.

Delivery: Includes a safety lid with retractable sheathed leads, UVT gel tray, two combs, one 12 well and one 20 well (1.5 mm thick).
Customers Who Bought This Also Bought