Moh's Hardness Collection
Catalog #: CA470313-110
Supplier: Avantor
Specifications
- Description:Mohs' Hardness Collection
- No. of Samples:9
- Cat. no.:CA470313-110
Specifications
About this item
Our most popular collection.
- Nine numbered samples
- Includes streak plate, nail, glass slide, and copper strip
All samples are pure crystalline forms to ensure accurate test results. You can also perform other physical properties tests with the included materials. The set is housed in a compartmented collection box, and an identification list is included.
Customers Who Bought This Also Bought