Interchangeable Glass Syringe with Luer Lock Tip, Ace Glass Incorporated
Catalog #: 76312-826
Supplier: Ace Glass
Specifications
- Capacity:10 ml
- Description:Interchangeable Syringe with Luer Lock Tip
- Material:Borosilicate Glass
- Cat. no.:76312-826
- Graduations:1/5 ml
Specifications
About this item
Interchangeable syringe featuring a Luer lock tip, permanently fused graduations, and a colored plunger end for easy and accurate dosage measurement.
- Made of high-quality borosilicate glass
- Reinforced flanges that are flat on two sides to prevent rolling
- Marked for non-medical use; no medical license required
Manufactured to ensure the perfect mating of barrels and plungers, so any interchangeable plunger of a particular size will fit any interchangeable barrel of the same size.
Customers Who Bought This Also Bought