Order Entry
ContactUsLinkComponent
Water-In-Methanol 5.0 Hydranal 1 L
Auxilary Reagents for Karl Fischer Titration, HYDRANAL™, Honeywell Research Chemicals
Catalog #: BJ-34802-1L
CAS Number:  
Auxilary Reagents for Karl Fischer Titration, HYDRANAL™, Honeywell Research Chemicals
Catalog #: BJ-34802-1L
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Packaging:
    Glass Bottle
  • Description:
    HYDRANAL™ Water-in-Methanol 5.0, Reagent for volumetric back titration, water content 5.00 mg/mL
  • Size:
    1 x 1 L
  • Cat. no.:
    BJ-34802-1L

Specifications

About this item

Karl Fischer titration is applied to multifarious substances. The nuances in sample properties influence the Karl Fischer titration differently. There are a number of ways to adjust the working conditions.

Solubilizers: In special cases the addition of solubilizer is required in order to enable a direct titration of the sample and avoid complicated and error-prone pre-dissolution and pre-extraction steps.

Buffers: The Karl Fischer reaction is pH dependant, with pH 5-7.5 being the ideal range. Strongly acidic samples slow the reaction and must be neutralized without inducing an alkaline reaction of the working medium prior to starting the titration. Strong bases can increase the pH of the working solution if the basicity exceeds the buffering capacity of the reagent. A titration end point will not be reached. Strong bases also must be neutralized prior to starting the titration.

Drying agents: Special drying agents are suitable to hold the near environment of the Karl Fischer equipment on a low water level or to dry
carrier gases in case of oven technique

HYDRANAL-Moisture Test Kit is for rough measurements without a titrator, special test kits for visual water determination according to Karl Fischer can be used.
The set contains syringes, titration vessel and reagents: 2 x 500 mL Hydranal-Solvent E (34730), 100 mL Hydranal-Titrant 5 E (34732) and 100 mL Hydranal-Standard 5.0 (34813). Refills can be ordered separately.

Customers Who Bought This Also Bought