VWR® Pierceable, Silicone Microplate Sealing Mats
Catalog #: 76311-638
Supplier: VWR International
Specifications
- Description:Microplate Sealing Mats for use with 96-Well Deep Well Plates with Square Holes
- Cat. no.:76311-638
Specifications
About this item
Suitable for sealing microplates in incubation and storage applications, silicone sealing mats provide sample protection while minimizing evaporation in a wide range of temperatures.
- Pierceable for easy penetration; self-healing
- Chemically resistant, research-grade silicone
- Contains no adhesives or extractables to contaminate samples
- Printed alpha numeric grid for easy reference
- Eliminates cross-contamination between wells
- Sample access via pipettor, syringe, or probe
- Airtight closure helps minimize sample evaporation
- Resists coring and tearing
Silicone microplate sealing mats are suitable for high-throughput applications and high-temperature storage. All mats are reusable; autoclave and follow with 10% bleach wash and ethanol rinse.
Certifications: Certified Rnase-, Dnase-, DNA- and Pyrogen-free.
Customers Who Bought This Also Bought