ISO17025 CALIBRATION CERTIFICATE 3PT
Catalog #: 76310-484
Supplier: ELPRO SERVICES, INC.
Customers Who Bought This Also Bought
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
$214.30 to $2,349.04
Sodium carbonate, anhydrous ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
$129.84 to $1,081.96
Copper(II) sulfate pentahydrate 98.0-102.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
$105.35 to $893.56
Reagent Reservoirs, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$277.83 to $277.83