Laboratory Aprons
Catalog #: 470311-692
Supplier: United Scientific Supplies
Specifications
- Garment application:Chemical protection,General Purpose,Laboratory
- Garment fit:Regular
- Gender:Unisex
- L×W:42×27"
- Fabric:PVC-coated fabric
- Cat. no.:470311-692
Specifications
About this item
Protective laboratory apron for multipurpose use.
- Flexible
- Chemical resistant
- Apron includes cloth ties at the neck and waist
These aprons are made of a PVC-coated fabric which stays flexible. There is reinforcement at the points of strain.
Customers Who Bought This Also Bought