Vial/Screw Cap Rsa-Pro Glass Ptfe PK100
Catalog #: 76304-508
Supplier: MicroSolv Technology Corp
Customers Who Bought This Also Bought
VWR® Prefilled Histology Containers, 10% Neutral Buffered Formalin (NBF)
Supplier: VWR International
$587.19
Diethyl ether ≥98% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
$775.79 to $2,413.44
Sodium hydroxide 0.25 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$184.00 to $683.98
Sulfuric acid ≥95%, BAKER INSTRA-ANALYZED® for trace metal analysis low Selenium, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$483.15 to $1,605.80