ContactUsLinkComponent
Vesphene Iii Disinfectant St 2 Oz CS144
Vesphene® III st Disinfectant Cleaners, Steris
Catalog #: 76304-176
Supplier:  Steris
Vesphene Iii Disinfectant St 2 Oz CS144
Vesphene® III st Disinfectant Cleaners, Steris
Catalog #: 76304-176
Supplier:  Steris
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    59.14 ml (2 oz.)
  • Packaging:
    Pouches
Specifications
About this item
These alkaline phenolic cleaners are specifically formulated for use on hard, nonporous surfaces.
  • Efficacy - Phenolic active ingredients provide broad spectrum antimicrobial efficacy.
  • Convenience - Gamma irradiated and double bagged for easy introduction into your aseptic areas.
  • Flexibility - Ideal for use in a contamination control rotation.
  • Reliability - Complete lot traceability.
  • Assurance - Each lot documented by Certificate of Analysis.
Vesphene® III st have dilution rate of 0.8% v/v (1:128) and is ideal for use in a disinfectant rotation program with acidic LpH® disinfectant cleaner.
The use-dilution can be applied manually or sprayed.
Customers Who Bought This Also Bought