Order Entry
ContactUsLinkComponent
Funnel Powder Glass 75Mm
Glass Powder Funnels
Catalog #: 470310-696
Supplier:  Wards
Funnel Powder Glass 75Mm
Glass Powder Funnels
Catalog #: 470310-696
Supplier:  Wards

Specifications

  • Ø:
    75 mm
  • Cat. no.:
    470310-696

Specifications

About this item

These funnels are available in a variety of different diameter openings.

  • Borosilicate glass
  • Plain 60° angle
  • Fast and efficient

When pouring large particles into reaction vessels and beakers, powder funnels allow a clean and quick procedure. Powder bridging is minimized by a parallel, short stem.

Customers Who Bought This Also Bought