Simple Distillation Apparatus
Catalog #: 470310-686
Supplier: Wards
Specifications
- Description:Simple Distillation Apparatus
- Cat. no.:470310-686
Specifications
About this item
This simple distillation apparatus is made up of boroscilicate glass.
Delivery: Apparatus includes round bottom flask (500 ml), still head with thermometer support, Liebig condensor (300 mm), receiver adapter and 3×joint clip (19/26).
Customers Who Bought This Also Bought