Melting Point Capillary Tubes
Catalog #: 470310-684
Supplier: Wards
Specifications
- Description:1 Open End
- Cat. no.:470310-684
Specifications
About this item
Borosilicate glass capillary melting tubes are 4" in length and have a 0.05" outer diameter capillary tubes that are particularly useful for determining the melting point of various substances.
The 100 capillary tubes come in a cylindrical container that has a hole in the lid for easily obtaining one tube at a time.
Customers Who Bought This Also Bought