Order Entry
ContactUsLinkComponent
Support Stand Rod Stamped Steel 5Inx8In
Stamped Steel Support Stands with Rods, United Scientific Supplies
Catalog #: 76293-618
Support Stand Rod Stamped Steel 5Inx8In
Stamped Steel Support Stands with Rods, United Scientific Supplies
Catalog #: 76293-618

Specifications

  • Cat. no.:
    76293-618
  • Base Dimensions:
    5×8"
  • Rod Diameter:
    ⅜"
  • Rod Height:
    20"

Specifications

About this item

Stamped steel bases, with black enamel finish.

  • Zinc plated steel rod

Sturdy steel base.

Ordering information: Bases and Rods are also available individually.
Delivery: Includes a removable, zinc-plated steel rod.
Customers Who Bought This Also Bought