Plastic Clamps for Jointed Glassware
Catalog #: 76293-448
Supplier: United Scientific Supplies
Specifications
- Color:Green
- Description:Clamp for jointed glassware
- Material:PP
- ST Joint Size:24/40
Specifications
About this item
Hold jointed sockets securely.
- Manufactured from polypropylene
- Outstanding chemical resistance
- Maximum operating temperature of 135 °C
- Autoclavable
Plastic conical joint clips are designed to securely hold joint size glassware together. The clamps are non-corrosive, and will not scratch glassware.
Customers Who Bought This Also Bought