Cast Aluminum Clamp Holder, United Scientific Supplies
Catalog #: 76293-416
Supplier: United Scientific Supplies
Specifications
- Description:Cast Aluminum Clamp Holder
- Cat. no.:76293-416
Specifications
About this item
Easily connect items to a support rod.
- Easy to adjust thumbscrews
- Fits rods up to 19 mm
Cast iron body with brass thumb screws.
Customers Who Bought This Also Bought