ContactUsLinkComponent
Hand Vacuum Pump Only For Filtering Kit
Hand Vacuum Pump, United Scientific Supplies
Catalog #: 76293-306
Hand Vacuum Pump Only For Filtering Kit
Hand Vacuum Pump, United Scientific Supplies
Catalog #: 76293-306
Specifications
  • Description:
    Hand Vacuum Pump
  • Cat. no.:
    76293-306
Specifications
About this item
Good quality, economical hand operated air/vacuum pump is easy to operate.
  • Recommended for general laboratory use
  • Use for applications that do not require high or precise vacuums
Each squeeze of the pump delivers approximately 15 ml, and can be used to create vacuum of 15 mm Hg, or air pressure of about 2 atm (standard atmosphere). Pump features a gauge along with a removable cap and elastic valve to provide quick vacuum release without dismantling the pump from the line.
Customers Who Bought This Also Bought