Rubber Stopper for Serum Bottles and Vials, Ace Glass Incorporated
Catalog #: 76291-836
Supplier: Ace Glass
Specifications
- Description:Snap-On Rubber Stopper
- Cat. no.:76291-836
- I.D.×O.D.:13×20 mm
Specifications
About this item
Gray, thin, flange-style chlorobutyl rubber stopper fits all 20 mm serum bottles and serum vials.
Customers Who Bought This Also Bought