


Nitric acid 67 - 70% (w/w), BAKER INSTRA-ANALYZED® Plus for trace metal analysis, for ICP, J.T.Baker®
Catalog # CAJT9368-5
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
MOC: Non-RegulatedCertificates
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Poly bottle
- Size:2.5 L
- Cat. no.:CAJT9368-5
Specifications
Specification Test Results
- Color (APHA):≤10
- Assay (HNO₃)(w/w):67 - 70 %
- Chloride (Cl):≤0.2 ppm
- Total Phosphorus (P):≤0.01 ppm
- Total Sulphur (S):≤0.3 ppm
- Trace Impurities - Aluminum (Al):≤1 ppb
- Trace Impurities - Antimony (Sb):≤0.5 ppb
- Trace Impurities - Arsenic (As):≤0.5 ppb
- Trace Impurities - Barium (Ba):≤0.1 ppb
- Trace Impurities - Beryllium (Be):≤0.1 ppb
- Trace Impurities - Bismuth (Bi):≤0.1 ppb
- Trace Impurities - Cadmium (Cd):≤0.5 ppb
- Trace Impurities - Calcium (Ca):≤1 ppb
- Trace Impurities - Cerium (Ce):≤0.1 ppb
- Trace Impurities - Cesium (Cs):≤0.1 ppb
- Trace Impurities - Cobalt (Co):≤0.1 ppb
- Trace Impurities - Copper (Cu):≤0.5 ppb
- Trace Impurities - Dysprosium (Dy):≤0.1 ppb
- Trace Impurities - Erbium (Er):≤0.1 ppb
- Trace Impurities - Europium (Eu):≤0.1 ppb
- Trace Impurities - Gadolinium (Gd):≤0.1 ppb
- Trace Impurities - Gallium (Ga):≤0.1 ppb
- Trace Impurities - Germanium (Ge):≤0.1 ppb
- Trace Impurities - Gold (Au):≤0.1 ppb
- Trace Impurities - Hafnium (Hf):≤0.1 ppb
- Trace Impurities - Holmium (Ho):≤0.1 ppb
- Trace Impurities - Indium (In):≤0.1 ppb
- Trace Impurities - Iron (Fe):≤1 ppb
- Trace Impurities - Lanthanum (La):≤0.1 ppb
- Trace Impurities - Lead (Pb):≤0.1 ppb
- Trace Impurities - Lithium (Li):≤0.1 ppb
- Trace Impurities - Lutetium (Lu):≤0.1 ppb
- Trace Impurities - Magnesium (Mg):≤1 ppb
- Trace Impurities - Manganese (Mn):≤0.1 ppb
- Trace Impurities - Mercury (Hg):≤0.1 ppb
- Trace Impurities - Molybdenum (Mo):≤0.1 ppb
- Trace Impurities - Neodymium (Nd):≤0.1 ppb
- Trace Impurities - Nickel (Ni):≤0.5 ppb
- Trace Impurities - Niobium (Nb):≤0.1 ppb
- Trace Impurities - Palladium (Pd):≤0.5 ppb
- Trace Impurities - Platinum (Pt):≤0.5 ppb
- Trace Impurities - Potassium (K):≤1 ppb
- Trace Impurities - Praseodymium (Pr):≤0.1 ppb
- Trace Impurities - Rhenium (Re):≤0.1 ppb
- Trace Impurities - Rhodium (Rh):≤0.5 ppb
- Trace Impurities - Rubidium (Rb):≤0.1 ppb
- Trace Impurities - Ruthenium (Ru):≤0.5 ppb
- Trace Impurities - Samarium (Sm):≤0.1 ppb
- Trace Impurities - Scandium (Sc):≤0.1 ppb
- Trace Impurities - Selenium (Se):≤1 ppb
- Trace Impurities - Silver (Ag):≤0.1 ppb
- Trace Impurities - Sodium (Na):≤1 ppb
- Trace Impurities - Strontium (Sr):≤0.1 ppb
- Trace Impurities - Tellurium (Te):≤0.1 ppb
- Trace Impurities - Terbium (Tb):≤0.1 ppb
- Trace Impurities - Thallium (Tl):≤0.1 ppb
- Trace Impurities - Thorium (Th):≤0.1 ppb
- Trace Impurities - Thulium (Tm):≤0.1 ppb
- Trace Impurities - Tin (Sn):≤0.5 ppb
- Trace Impurities - Titanium (Ti):≤0.5 ppb
- Trace Impurities - Tungsten (W):≤0.1 ppb
- Trace Impurities - Uranium (U):≤0.1 ppb
- Trace Impurities - Vanadium (V):≤0.5 ppb
- Trace Impurities - Ytterbium (Yb):≤0.1 ppb
- Trace Impurities - Yttrium (Y):≤0.1 ppb
- Trace Impurities - Zinc (Zn):≤0.5 ppb
- Trace Impurities - Zirconium (Zr):≤0.1 ppb
Specification Test Results
About this item
:
:63.01
Merck Index:13,06608
Boiling Pt:120.5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
:Management of Change (MOC) category = N
:GHS03
:GHS05
:GHS06
Frequently Bought Together