Acetone ≥99.5% (by GC), Finyte® for microelectronic, J.T.Baker®
Catalog #: CAJT5008-5
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 67-64-1
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly bottle, pressure bottom
- Size:4 L
Specifications
Specification Test Results
- For Microelectronic Use:
- Assay (by GC):≥99.5 %
- Color (APHA):≤10
- Acidity (µeq/g):≤0.3
- Aldehydes (as HCHO):≤20000 ppb
- Alkalinity (µeq/g):≤0.5
- Isopropyl Alcohol (CH₃CHOHCH₃)(BY WT):≤0.05 %
- Methanol (CH₃OH)(BY WT):≤0.05 %
- Sulfate (SO₄), For Information Only:
- Residue after Evaporation:≤3000.0 ppb
- Water (H₂O):≤0.50 %
- Solubility in H₂O:Passes Test
- Chloride (Cl):≤200.0 ppb
- Phosphate (PO₄):≤50.0 ppb
- Trace Impurities - Aluminum (Al):≤10.0 ppb
- Trace Impurities - Antimony (Sb):≤5.0 ppb
- Trace Impurities - Arsenic (As):≤5.0 ppb
- Trace Impurities - Barium (Ba):≤10.0 ppb
- Trace Impurities - Beryllium (Be):≤10.0 ppb
- Trace Impurities - Bismuth (Bi):≤10.0 ppb
- Trace Impurities - Boron (B):≤10.0 ppb
- Trace Impurities - Cadmium (Cd):≤10.0 ppb
- Trace Impurities - Calcium (Ca):≤10.0 ppb
- Trace Impurities - Chromium (Cr):≤10.0 ppb
- Trace Impurities - Cobalt (Co):≤10.0 ppb
- Trace Impurities - Copper (Cu):≤10.0 ppb
- Trace Impurities - Gallium (Ga):≤10.0 ppb
- Trace Impurities - Germanium (Ge):≤10.0 ppb
- Trace Impurities - Gold (Au):≤10.0 ppb
- Trace Impurities - Iron (Fe):≤10.0 ppb
- Trace Impurities - Lead (Pb):≤10.0 ppb
- Trace Impurities - Lithium (Li):≤10.0 ppb
- Trace Impurities - Magnesium (Mg):≤10.0 ppb
- Trace Impurities - Manganese (Mn):≤10.0 ppb
- Trace Impurities - Molybdenum (Mo):≤10.0 ppb
- Trace Impurities - Nickel (Ni):≤10.0 ppb
- Trace Impurities - Niobium (Nb):≤10.0 ppb
- Trace Impurities - Potassium (K):≤10.0 ppb
- Trace Impurities - Silicon:≤10.0 ppb
- Trace Impurities - Silver (Ag):≤10.0 ppb
- Trace Impurities - Sodium (Na):≤10.0 ppb
- Trace Impurities - Strontium (Sr):≤10.0 ppb
- Trace Impurities - Tantalum (Ta):≤10.0 ppb
- Trace Impurities - Thallium (Tl):≤10.0 ppb
- Trace Impurities - Tin (Sn):≤10.0 ppb
- Trace Impurities - Titanium (Ti):≤10.0 ppb
- Trace Impurities - Vanadium (V):≤10.0 ppb
- Trace Impurities - Zinc (Zn):≤10.0 ppb
- Trace Impurities - Zirconium (Zr):≤10.0 ppb
- Particle Count - 0.5 µm and greater:≤50 par/ml
- Particle Count - 1.0 µm and greater:≤8 par/ml
- J.T.Baker®, Acetone ≥99.5% (by GC), Finyte® for microelectronic:
Specification Test Results
About this item
Synonyms: 2-Propanone,DMK,Propanone
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Management of Change (MOC) category = R
Flash Pt:<–20 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 56.2 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -95.4 °C|Melting Pt: –95.4 °C
:Flash Pt:
:Flash Pt: <–20 °C (closed cup)
:M.W. 58.08 g/mol
Merck Index:13,00067
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
REACH:01-2119471330-49
:3
:II
:<–20 °C
:GHS02
:GHS07
:GHS08
Customers Who Bought This Also Bought