Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Catalog #: CAJT4102-F7
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 77-86-1
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:HDPE drum, Flowmor™ package
- Size:12 kg
Specifications
Specification Test Results
- Meets JPC (1997) Chemical Specifications:
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- Meets U.S.P Requirements:
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (C₄H₁₁NO₃):99.0 - 101.0 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Identification C:Passes Test
- USP - pH (1 in 20):10.0 - 11.5
- USP - Loss on Drying:≤1.0 %
- USP - Residue on Ignition:≤0.1 %
- USP - Melting Range(Lower):168 - 172 °C
- USP - Melting Range(Upper):168 - 172 °C
- EP/BP - Assay (C₄H₁₁NO₃) (dried basis):99.0 - 100.5 %
- EP/BP - Identification B (melting point):168 - 174 °C
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - pH:10.0 - 11.5
- EP/BP - Related Substances:≤1.0 %
- EP/BP - Chloride (Cl):≤100 ppm
- EP/BP - Iron (Fe):≤10 ppm
- EP/BP - Loss on Drying at 105°C:≤0.5 %
- EP/BP - Ash (sulfated):≤0.1 %
- JPC 1997 - Assay (C₄H₁₁NO₃) (dried basis):≥99.0 %
- JPC 1997 - Identification A:Passes Test
- JPC 1997 - Identification B:Passes Test
- JPC 1997 - pH:10.3 - 10.7
- JPC 1997 - Melting Point:168 - 172 °C
- JP - Clarity and Color of Solution:Passes Test
- JPC 1997 - Heavy Metals (as Pb):≤8 ppm
- JPC 1997 - Arsenic (As):≤1.6 ppm
- JPC 1997 - Loss on Drying:≤0.5 %
- JPC 1997 - Residue on Ignition:≤0.10 %
- Assay (C₄H₁₁NO₃) (dried basis):≥99.9 %
- Absorbance of a 1M Solution - 260 nm:≤0.06
- Absorbance of a 1M Solution - 280 nm:≤0.06
- Absorbance of a 1M Solution - 400 nm:≤0.01
- Endotoxin Concentration (EU/g):≤2.5
- Iron (Fe):≤1 ppm
- Lead (Pb):≤1 ppm
- Ultrapure Bioreagent Specifications:
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Manufactured exclusively for Avantor Performance Materials, LLC., by BioSpectra, Inc., Stroudsburg, PA:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial:
Specification Test Results
About this item
Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris),Tromethamine,Tris(hydroxymethyl)methylamine,2-Amino-2-(hydroxymethyl)propane-1,3-diol,2-Amino-2-(hydroxymethyl)-1,3-propanediol,Trizma(R) base,TRIS base
Boiling Pt:357 °C (1013 hPa)
Melting Pt:169…172 °C
Density:1.328 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-101,0
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 357 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 169 to 172 °C|Melting Pt: 169…172 °C
:Flash Pt:
:Flash Pt:
:M.W. 121.14 g/mol
Merck Index:12,09902
:Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Storage Temperature:Ambient
EINECS:201-064-4
:GHS07
Customers Who Bought This Also Bought