Orthophosphoric acid ≥85.0% (by acidimetry), AR® ACS, Macron Fine Chemicals™
Catalog #: CAJT2796-18
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 7664-38-2
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, clear, Mall
- Size:500 ml
- Cat. no.:CAJT2796-18
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (as H₃PO₄) (by acidimetry):≥85.0 %
- ACS - Antimony (Sb):≤15.000 ppm
- ACS - Arsenic (As):≤1.000 ppm
- ACS - Calcium (Ca):≤0.002 %
- ACS - Chloride (Cl):≤3 ppm
- ACS - Color (APHA):≤10
- ACS - Heavy Metals (as Pb):≤10 ppm
- ACS - Insoluble Matter:≤0.001 %
- ACS - Iron (Fe):≤6 ppm
- ACS - Magnesium (Mg):≤0.002 %
- ACS - Manganese (Mn):≤0.500 ppm
- ACS - Nitrate (NO₃):≤5 ppm
- ACS - Potassium (K):≤6.000 ppm
- ACS - Reducing Substances (mL):≤1.0
- ACS - Sodium (Na):≤10.000 ppm
- ACS - Sulfate (SO₄):≤0.001 %
- ACS - Volatile Acids (as CH₃COOH):≤0.001 %
- Product Information (not specifications)::
- Appearance (clear, colorless liquid):
- Macron Fine Chemicals™, Orthophosphoric acid ≥85.0% (by acidimetry), AR® ACS:
Specification Test Results
About this item
Synonyms: Phosphoric acid
Melting Pt:28 °C
Density:1.71…1.87 g/cm³ (25 °C)
ADR:8,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:85,0
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 28 °C
:Flash Pt:
:Flash Pt:
:M.W. 98 g/mol
EINECS:231-633-2
:158
UN:1805
:8
:III
:GHS05
:GHS07
Storage Temperature:Ambient
:M.O.C. Category N
:12,07500
:14,07344
Customers Who Bought This Also Bought