ContactUsLinkComponent
Syringe Exel 1Cc X 30G X 5/16In BX100
Exel International® Insulin Syringes with Fixed Needles, Air-Tite Products
Catalog #: 76290-416
Supplier:  Air-Tite
Syringe Exel 1Cc X 30G X 5/16In BX100
Exel International® Insulin Syringes with Fixed Needles, Air-Tite Products
Catalog #: 76290-416
Supplier:  Air-Tite
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Length:
    5/16"
  • Volume:
    1 ml
  • Description:
    Syringes with Fixed Needles
  • Sterility:
    Sterile
  • Packaging:
    100/box
  • For Use With:
    Human, Laboratory, and Veterinary Use
  • Gauge:
    30
  • Type:
    Insulin
Specifications
About this item
These EXEL International Insulin Syringes with Fixed Needle are high quality and cost-efficient.
  • Sterile
  • Latex free
  • Non pyrogenic
  • Non toxic
  • Low deadspace
The insulin syringes design includes bold printing, clear graduations and siliconized gaskets for smooth consistent plunger motion and a positive plunger stop. The auto ground needle tip provides for greater comfort. Reduced dead space in this design makes for reduced Insulin waste.
Certifications: Meets ISO Standards, CE registered.
Packaging: Sterile and wrapped packs of 10.
Customers Who Bought This Also Bought