Order Entry
ContactUsLinkComponent
Tubing Connection Port 1/4-28 Or 2.5Mm
Vaplock™ Tubing Connections, Cole-Parmer
Catalog #: 76287-578
Supplier:  Argos Technologies
Tubing Connection Port 1/4-28 Or 2.5Mm
Vaplock™ Tubing Connections, Cole-Parmer
Catalog #: 76287-578
Supplier:  Argos Technologies

Specifications

  • Color:
    Orange
  • Description:
    Tubing Connection; 2.5 mm O.D. Tubing×1/4-28, Orange PP Fitting, Natural Tefzel Ferrule
  • Cat. no.:
    76287-578
  • Fits Tubing O.D.:
    2.5 mm

Specifications

About this item

Liquid-tight tubing connections for 1/4-28 and 5/16-24 ports.

Compression fittings designed for liquid-tight connection of semi-rigid tubing to flat-bottom 1/4-28 and 5/16-24 port threads, for 1/16" to 3/16" O.D. tubing. Each connection is comprised of a two-piece nut and ferrule system, useful for pressurized applications under 500 psi. Select based on tubing outer diameter. Fittings are color-coded by tubing size for easy identification. These bottom-sealing UNF fittings do not require PTFE tape.

Use with 1/16" to 3/16" O.D. tubing.

Customers Who Bought This Also Bought