ContactUsLinkComponent
PfuUltra High-Fidelity DNA Polymerase 100 unit
PfuUltra High-Fidelity DNA Polymerase, Agilent Technologies
Catalog #: CAST600380
CAS Number:  
PfuUltra High-Fidelity DNA Polymerase 100 unit
PfuUltra High-Fidelity DNA Polymerase, Agilent Technologies
Catalog #: CAST600380
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    PfuUltra High-Fidelity DNA Polymerase
  • Size:
    100 unit
  • Cat. no.:
    CAST600380
Specifications
About this item
PfuUltra High-Fidelity DNA Polymerase AD features a reformulated buffer system for increased economy with the same high performance as the original PfuUltra DNA Polymerase for robust, ultra-high-fidelity PCR.
PfuUltra Hotstart DNA Polymerase features a genetically engineered mutant of Pfu DNA Polymerase and the ArchaeMaxx Polymerase Enhancing Factor and is formulated for hot start activity.
PfuUltra Hotstart DNA Polymerase AD features a reformulated buffer system and novel hot start mechanism for increased economy with the same high performance as the original PfuUltra Hotstart DNA Polymerase.
PfuUltra Hotstart PCR Master Mix is a 2X formulation of PfuUltra Hotstart DNA Polymerase, an optimized PCR reaction buffer, magnesium, and dNTPs.
Customers Who Bought This Also Bought