ContactUsLinkComponent
MycoSensor QPCR Assay Kit 50 reactions
MycoSensor qPCR Assay Kits, Agilent Technologies
Catalog #: CAST302107
CAS Number:  
MycoSensor QPCR Assay Kit 50 reactions
MycoSensor qPCR Assay Kits, Agilent Technologies
Catalog #: CAST302107
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    MycoSensor qPCR assay kit
  • Size:
    50 reactions
  • Cat. no.:
    CAST302107
  • Details:
    All the reagents you need for amplification and fluorescence detection, internal controls, DNA purification reagents.
Specifications
About this item
The MycoSensor PCR and qPCR assay kits detect cultured Mycoplasma infection by real-time qPCR or PCR.
  • Detects Mycoplasma infection of cell cultures by qPCR or PCR in less than two hours
  • Specific detection of the eight most common Mycoplasma species
  • SYBR Green dye detection
The MycoSensor PCR and qPCR assay kits detect Mycoplasma infection of cell cultures by real-time quantitative PCR (qPCR) or PCR in less than two hours. The easy-to-use system can specifically detect the eight most common Mycoplasma species.
The system uses SYBR Green dye detection. The kit includes a primer mix that amplifies the eight most common Mycoplasma species that infect cell cultures. The MycoSensor qPCR assay kit primer mix can detect Mycoplasma infections in cell cultures in less than two hours.
Customers Who Bought This Also Bought